Assay Details
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112712.1 | NP_001106183.1 | 2 - 3 | 160 | 104 | |
NM_001112713.1 | NP_001106184.1 | 2 - 3 | 160 | 104 | ||
NM_031657.3 | NP_113845.2 | 2 - 3 | 160 | 104 | ||
XM_006253653.2 | XP_006253715.1 | 2 - 3 | 269 | 104 | ||
XM_008771504.1 | XP_008769726.1 | 2 - 3 | 152 | 104 | ||
GenBank mRNA | CV795758.1 | - | 6 - 7 | 104 | ||
EX488847.1 | - | 2 - 3 | 320 | 104 | ||
Y09365.1 | - | 2 - 3 | 188 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006253653.2, XM_008771504.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
