Assay Details
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024795.1 | NP_001019966.1 | 4 - 5 | 678 | 89 | |
XM_008771498.1 | XP_008769720.1 | 4 - 5 | 2194 | 89 | ||
GenBank mRNA | BC098030.1 | - | 4 - 5 | 678 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
