Assay Details
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235892.2 | XP_006235954.1 | 7 - 8 | 1381 | 124 | |
XM_006235894.2 | XP_006235956.1 | 7 - 8 | 1368 | 124 | ||
XM_006235895.1 | XP_006235957.1 | 7 - 8 | 1365 | 124 | ||
XM_006235896.2 | XP_006235958.1 | 7 - 8 | 1382 | 124 | ||
GenBank mRNA | FQ215763.1 | - | 8 - 9 | 1243 | 124 | |
FQ228972.1 | - | 7 - 8 | 1321 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
