Assay Details
Gene Symbol: LOC499971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024317.2 | NP_001019488.2 | 10 - 11 | 1342 | 69 | |
XM_008762655.1 | XP_008760877.1 | 9 - 10 | 995 | 69 | ||
XM_008762656.1 | XP_008760878.1 | 8 - 9 | 886 | 69 | ||
GenBank mRNA | BC079392.1 | - | 3 - 4 | 278 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
