Assay Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109666.1 | NP_001103136.1 | 11 - 12 | 1576 | 98 | |
XM_006235929.2 | XP_006235991.1 | 12 - 13 | 1546 | 98 | ||
XM_006235930.2 | XP_006235992.1 | 12 - 13 | 1708 | 98 | ||
XM_006235931.2 | XP_006235993.1 | 12 - 13 | 1543 | 98 | ||
XM_006235932.2 | XP_006235994.1 | 12 - 13 | 1537 | 98 | ||
XM_006235933.2 | XP_006235995.1 | 12 - 13 | 1534 | 98 | ||
XM_006235934.2 | XP_006235996.1 | 11 - 12 | 1456 | 98 | ||
GenBank mRNA | BC129119.1 | - | 11 - 12 | 1443 | 7115823 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
