Assay Details
Gene Symbol: Atg9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191968.1 | NP_001178897.1 | 4 - 5 | 826 | 66 | |
XM_006235960.2 | XP_006236022.1 | 4 - 5 | 826 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
