Assay Details
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 10 - 11 | 1070 | 90 | |
GenBank mRNA | D50694.1 | - | 10 - 11 | 1070 | 90 | |
FQ213265.1 | - | 7 - 8 | 873 | 90 | ||
FQ215767.1 | - | 7 - 8 | 873 | 90 | ||
FQ225383.1 | - | 7 - 8 | 873 | 90 | ||
FQ230973.1 | - | 10 - 11 | 1128 | 90 | ||
FQ234824.1 | - | 7 - 8 | 873 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
