Assay Details
Gene Symbol: Cops5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025695.1 | NP_001020866.1 | 5 - 6 | 773 | 146 | |
XM_006237758.2 | XP_006237820.1 | 5 - 6 | 713 | 146 | ||
GenBank mRNA | BC098736.1 | - | 5 - 6 | 773 | 7385252 | 146 |
FQ214411.1 | - | 5 - 6 | 696 | 146 | ||
FQ215612.1 | - | 5 - 6 | 696 | 146 | ||
FQ215908.1 | - | 5 - 6 | 696 | 146 | ||
FQ227775.1 | - | 5 - 6 | 697 | 146 | ||
FQ232542.1 | - | 5 - 6 | 696 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
