Assay Details
Gene Symbol: Ndufaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276441.1 | NP_001263370.1 | 3 - 4 | 426 | 101 | |
GenBank mRNA | FQ215404.1 | - | 3 - 4 | 426 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
