Assay Details
Gene Symbol: Ndufaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276441.1 | NP_001263370.1 | 1 - 2 | 205 | 76 | |
GenBank mRNA | FQ215404.1 | - | 1 - 2 | 205 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
