Assay Details
Gene Symbol: Ldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106009.1 | NP_001099479.1 | 4 - 5 | 757 | 157 | |
XM_008770199.1 | XP_008768421.1 | 4 - 5 | 758 | 157 | ||
XM_008770200.1 | XP_008768422.1 | 4 - 5 | 758 | 157 | ||
XM_008770201.1 | XP_008768423.1 | 4 - 5 | 758 | 157 | ||
XM_008770202.1 | XP_008768424.1 | 4 - 5 | 758 | 157 | ||
XM_008770203.1 | XP_008768425.1 | 4 - 5 | 758 | 157 | ||
GenBank mRNA | BC168881.1 | - | 4 - 5 | 642 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
