Assay Details
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022608.1 | NP_072130.1 | 5 - 6 | 449 | 89 | |
GenBank mRNA | AJ132352.1 | - | 5 - 6 | 464 | 89 | |
BC162038.1 | - | 5 - 6 | 449 | 7099625 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
