Assay Details
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 4 - 5 | 891 | 76 | |
XM_006251211.2 | XP_006251273.1 | 4 - 5 | 1094 | 76 | ||
XM_006251212.2 | XP_006251274.1 | 4 - 5 | 1094 | 76 | ||
XM_006251213.2 | XP_006251275.1 | 4 - 5 | 1260 | 76 | ||
XM_006251214.2 | XP_006251276.1 | 4 - 5 | 502 | 76 | ||
GenBank mRNA | BC081868.1 | - | 4 - 5 | 891 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
