Assay Details
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011912.1 | NP_001011912.1 | 6 - 7 | 1087 | 64 | |
XM_006251212.2 | XP_006251274.1 | 6 - 7 | 1290 | 64 | ||
GenBank mRNA | BC081868.1 | - | 6 - 7 | 1087 | 7096894 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
