Assay Details
Gene Symbol: Lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169113.1 | NP_001162584.1 | 3 - 4 | 515 | 77 | |
GenBank mRNA | BC082020.1 | - | 3 - 4 | 483 | 77 | |
BC098947.1 | - | 3 - 4 | 480 | 77 | ||
FM055449.1 | - | 3 - 4 | 517 | 77 | ||
Z11994.1 | - | 475 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
