Assay Details
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011910.1 | NP_001011910.1 | 6 - 7 | 745 | 129 | |
GenBank mRNA | BC079381.1 | - | 6 - 7 | 745 | 129 | |
FQ218842.1 | - | 2 - 3 | 263 | 129 | ||
FQ235301.1 | - | 6 - 7 | 755 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
