Assay Details
Gene Symbol: Uvssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134558.1 | NP_001128030.1 | 12 - 13 | 2340 | 67 | |
XM_006251321.2 | XP_006251383.1 | 12 - 13 | 2359 | 67 | ||
XM_006251323.2 | XP_006251385.1 | 12 - 13 | 2075 | 67 | ||
XM_006251324.2 | XP_006251386.1 | 12 - 13 | 2006 | 67 | ||
XM_006251325.2 | XP_006251387.1 | 12 - 13 | 2133 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
