Assay Details
Gene Symbol: Uvssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134558.1 | NP_001128030.1 | 1 - 2 | 444 | 131 | |
XM_006251321.2 | XP_006251383.1 | 1 - 2 | 463 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134558.1, XM_006251321.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
