Assay Details
Gene Symbol: Uvssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134558.1 | NP_001128030.1 | 9 - 10 | 1925 | 114 | |
XM_006251321.2 | XP_006251383.1 | 9 - 10 | 1944 | 114 | ||
XM_006251323.2 | XP_006251385.1 | 9 - 10 | 1660 | 114 | ||
XM_006251324.2 | XP_006251386.1 | 9 - 10 | 1591 | 114 | ||
XM_006251325.2 | XP_006251387.1 | 9 - 10 | 1718 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
