Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270849.1 | NP_001257778.1 | 6 - 7 | 784 | 65 | |
NM_001270850.1 | NP_001257779.1 | 5 - 6 | 562 | 65 | ||
NM_012565.2 | NP_036697.1 | 5 - 6 | 685 | 65 | ||
XM_006251179.2 | XP_006251241.1 | 5 - 6 | 1530 | 65 | ||
GenBank mRNA | J04218.1 | - | 5 - 6 | 644 | 65 | |
KJ026953.1 | - | 5 - 6 | 577 | 65 | ||
M25807.1 | - | 5 - 6 | 562 | 65 | ||
X53588.1 | - | 5 - 6 | 668 | 65 | ||
X53589.1 | - | 6 - 7 | 784 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
