Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270849.1 | NP_001257778.1 | 7 - 8 | 888 | 85 | |
NM_001270850.1 | NP_001257779.1 | 6 - 7 | 666 | 85 | ||
NM_012565.2 | NP_036697.1 | 6 - 7 | 789 | 85 | ||
XM_006251179.2 | XP_006251241.1 | 6 - 7 | 1634 | 85 | ||
GenBank mRNA | J04218.1 | - | 6 - 7 | 748 | 85 | |
KJ026953.1 | - | 6 - 7 | 681 | 85 | ||
M25807.1 | - | 6 - 7 | 666 | 85 | ||
X53588.1 | - | 6 - 7 | 772 | 85 | ||
X53589.1 | - | 7 - 8 | 888 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
