Assay Details
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105750.2 | NP_001099220.2 | 10 - 11 | 1705 | 76 | |
XM_008770187.1 | XP_008768409.1 | 10 - 11 | 8347 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
