Assay Details
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105750.2 | NP_001099220.2 | 4 - 5 | 726 | 123 | |
XM_008770187.1 | XP_008768409.1 | 4 - 5 | 7368 | 123 | ||
GenBank mRNA | FQ231655.1 | - | 5 - 6 | 458 | 123 | |
FQ235297.1 | - | 5 - 6 | 458 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
