Assay Details
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105750.2 | NP_001099220.2 | 7 - 8 | 1194 | 77 | |
XM_008770187.1 | XP_008768409.1 | 7 - 8 | 7836 | 77 | ||
GenBank mRNA | FQ231655.1 | - | 8 - 9 | 926 | 77 | |
FQ235297.1 | - | 8 - 9 | 926 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
