Assay Details
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107238.1 | NP_001100708.1 | 3 - 4 | 744 | 74 | |
XM_006251444.2 | XP_006251506.1 | 4 - 5 | 582 | 74 | ||
GenBank mRNA | BC168726.1 | - | 4 - 5 | 554 | 74 | |
BC168735.1 | - | 4 - 5 | 554 | 74 | ||
BC168766.1 | - | 4 - 5 | 554 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
