Assay Details
Gene Symbol: Pes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044228.1 | NP_001037693.1 | 10 - 11 | 1179 | 112 | |
GenBank mRNA | BC105910.1 | - | 10 - 11 | 1179 | 7388101 | 112 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
