Assay Details
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012823.2 | NP_036955.2 | 12 - 13 | 1046 | 134 | |
XM_006250694.2 | XP_006250756.1 | 12 - 13 | 1410 | 134 | ||
XM_006250696.2 | XP_006250758.1 | 13 - 14 | 1166 | 134 | ||
GenBank mRNA | BC081856.1 | - | 11 - 12 | 958 | 134 | |
FQ232970.1 | - | 12 - 13 | 1039 | 134 | ||
M20559.1 | - | 12 - 13 | 994 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis
positive regulation of endothelial cell migration
hippocampus development
animal organ regeneration
defense response to bacterium
negative regulation of catalytic activity
neutrophil degranulation
positive regulation of angiogenesis
positive regulation of DNA metabolic process
positive regulation of DNA-binding transcription factor activity
response to glucocorticoid
response to growth factor
phospholipase inhibitor activity
calcium ion binding
calcium-dependent phospholipid binding
phospholipase A2 inhibitor activity
calcium-dependent protein binding
phagocytosis
positive regulation of endothelial cell migration
hippocampus development
animal organ regeneration
defense response to bacterium
negative regulation of catalytic activity
neutrophil degranulation
positive regulation of angiogenesis
positive regulation of DNA metabolic process
positive regulation of DNA-binding transcription factor activity
response to glucocorticoid
response to growth factor
phospholipase inhibitor activity
calcium ion binding
calcium-dependent phospholipid binding
phospholipase A2 inhibitor activity
calcium-dependent protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P116690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P325508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100665 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325508 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325508 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116690 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P116690 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P325508 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P301936 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11577 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P325508 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44893 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7503 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 99244 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10775639 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775643 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775640 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775641 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775644 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775642 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775638 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10775628 | - | - |
Rat Genome 230 2.0 Array | 1367974_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367974_at | - | - |
Rat Genome 230 2.0 Array | 1395313_PM_s_at | - | - |
Rat Genome 230 2.0 Array | 1380751_at | - | - |
Rat Genome 230 2.0 Array | 1367975_at | - | - |
Rat Genome 230 2.0 Array | 1395313_s_at | - | - |
Rat Genome 230 2.0 Array | 1367975_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380751_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367975_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395313_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1367975_at | - | - |
Affymetrix HT Rat Focus Array | 1367974_at | - | - |
Affymetrix HT Rat Focus Array | 1367974_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395313_s_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400010547.rn.1 | TC1400001444.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010546.rn.1 | TC1400001444.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010548.rn.1 | TC1400001444.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400010544.rn.1 | TC1400001444.rn.1 | RTA TC |
Rat Expression Set 230 | 1367975_at | - | - |
Rat Expression Set 230 | 1367974_PM_at | - | - |
Rat Expression Set 230 | 1380751_at | - | - |
Rat Expression Set 230 | 1367974_at | - | - |
Rat Expression Set 230 | 1367975_PM_at | - | - |
Rat Expression Set 230 | 1380751_PM_at | - | - |
Rat Expression Set 230 | 1395313_PM_s_at | - | - |
Rat Expression Set 230 | 1395313_s_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775628 | - | - |
Rat Genome U34 Set | rc_AI639072_at | - | - |
Rat Genome U34 Set | AA850219_at | - | - |
Rat Genome U34 Set | rc_AA945932_at | - | - |
Rat Genome U34 Set | rc_AA945932_g_at | - | - |
Rat Genome U34 Set | rc_AI179982_s_at | - | - |
Rat Genome U34 Set | rc_AI146189_s_at | - | - |
Rat Genome U34 Set | rc_AI179982_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22000040 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368619 | - | - |
RatRef-12 Expression BeadChip | ILMN_57519 | - | - |
