Assay Details
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108355.1 | NP_001101825.1 | 11 - 12 | 1588 | 111 | |
XM_006250708.2 | XP_006250770.1 | 11 - 12 | 1620 | 111 | ||
XM_006250709.2 | XP_006250771.1 | 11 - 12 | 1507 | 111 | ||
XM_008770042.1 | XP_008768264.1 | 10 - 11 | 4387 | 111 | ||
XM_008770043.1 | XP_008768265.1 | 10 - 11 | 1399 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
positive regulation of cell population proliferation
positive regulation of cytoplasmic mRNA processing body assembly
mRNA destabilization
RNA phosphodiester bond hydrolysis, exonucleolytic
poly(A)-specific ribonuclease activity
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
positive regulation of cell population proliferation
positive regulation of cytoplasmic mRNA processing body assembly
mRNA destabilization
RNA phosphodiester bond hydrolysis, exonucleolytic
poly(A)-specific ribonuclease activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P264901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55971 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P264901 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P264901 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P264901 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P264901 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P129900 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88066 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10771558 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771561 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771566 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771568 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771564 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771560 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771559 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771557 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771565 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771563 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771562 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10771556 | - | - |
Rat Genome 230 2.0 Array | 1393619_at | - | - |
Rat Genome 230 2.0 Array | 1392796_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392796_at | - | - |
Rat Genome 230 2.0 Array | 1393619_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393619_at | - | - |
Affymetrix HT Rat Focus Array | 1393619_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400001768.rn.1 | TC1400000221.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400001767.rn.1 | TC1400000221.rn.1 | RTA TC |
Rat Expression Set 230 | 1393619_PM_at | - | - |
Rat Expression Set 230 | 1392796_PM_at | - | - |
Rat Expression Set 230 | 1392796_at | - | - |
Rat Expression Set 230 | 1393619_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10771556 | - | - |
Rat Genome U34 Set | rc_AI045762_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20708360 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374682 | - | - |
