Assay Details
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108355.1 | NP_001101825.1 | 2 - 3 | 263 | 124 | |
XM_006250708.2 | XP_006250770.1 | 2 - 3 | 295 | 124 | ||
XM_006250709.2 | XP_006250771.1 | 2 - 3 | 182 | 124 | ||
XM_008770042.1 | XP_008768264.1 | 1 - 2 | 3062 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
