Assay Details
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108355.1 | NP_001101825.1 | 6 - 7 | 692 | 106 | |
XM_006250708.2 | XP_006250770.1 | 6 - 7 | 724 | 106 | ||
XM_006250709.2 | XP_006250771.1 | 6 - 7 | 611 | 106 | ||
XM_008770042.1 | XP_008768264.1 | 5 - 6 | 3491 | 106 | ||
XM_008770043.1 | XP_008768265.1 | 5 - 6 | 503 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
