Assay Details
Gene Symbol: Cnot6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108355.1 | NP_001101825.1 | 7 - 8 | 848 | 83 | |
XM_006250708.2 | XP_006250770.1 | 7 - 8 | 880 | 83 | ||
XM_006250709.2 | XP_006250771.1 | 7 - 8 | 767 | 83 | ||
XM_008770042.1 | XP_008768264.1 | 6 - 7 | 3647 | 83 | ||
XM_008770043.1 | XP_008768265.1 | 6 - 7 | 659 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
