Assay Details
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105997.2 | NP_001099467.1 | 4 - 5 | 505 | 93 | |
XM_008770009.1 | XP_008768231.1 | 5 - 6 | 872 | 93 | ||
GenBank mRNA | BC168772.1 | - | 4 - 5 | 505 | 6920150 | 93 |
FQ217255.1 | - | 4 - 5 | 507 | 93 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750509.3 | XP_003750557.1 | 4 - 5 | 499 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
