Assay Details
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105997.2 | NP_001099467.1 | 7 - 8 | 797 | 123 | |
XM_008770009.1 | XP_008768231.1 | 8 - 9 | 1164 | 123 | ||
GenBank mRNA | BC168772.1 | - | 7 - 8 | 797 | 123 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750509.3 | XP_003750557.1 | 7 - 8 | 791 | 123 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1010226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22699 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1010226 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1010226 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1010226 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1010226 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14635 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98066 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10775726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775720 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10775719 | - | - |
Rat Genome 230 2.0 Array | 1389125_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389125_at | - | - |
Affymetrix HT Rat Focus Array | 1389125_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389125_at | - | - |
Rat Expression Set 230 | 1389125_PM_at | - | - |
Rat Expression Set 230 | 1389125_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10775719 | - | - |
Rat Genome U34 Set | rc_AA818414_at | - | - |
Rat Genome U34 Set | rc_AI176979_at | - | - |
Rat Genome U34 Set | rc_AI180110_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22391407 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363224 | - | - |
