Assay Details
Gene Symbol: Nxpe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109435.1 | NP_001102905.1 | 2 - 3 | 576 | 79 | |
XM_006248252.2 | XP_006248314.1 | 2 - 3 | 402 | 79 | ||
XM_006248253.2 | XP_006248315.1 | 2 - 3 | 413 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
