Assay Details
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079938.1 | NP_001073407.1 | 10 - 11 | 2242 | 105 | |
XM_008768634.1 | XP_008766856.1 | 10 - 11 | 2134 | 105 | ||
XM_008768635.1 | XP_008766857.1 | 10 - 11 | 2329 | 105 | ||
XM_008768636.1 | XP_008766858.1 | 11 - 12 | 2443 | 105 | ||
XM_008768637.1 | XP_008766859.1 | 11 - 12 | 2248 | 105 | ||
XM_008768638.1 | XP_008766860.1 | 10 - 11 | 2134 | 105 | ||
XM_008768639.1 | XP_008766861.1 | 10 - 11 | 2329 | 105 | ||
XM_008768640.1 | XP_008766862.1 | 10 - 11 | 2020 | 105 | ||
XM_008768641.1 | XP_008766863.1 | 10 - 11 | 2134 | 105 | ||
GenBank mRNA | BC129113.1 | - | 10 - 11 | 2242 | 105 |
Gene Symbol: LOC100912484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751043.3 | XP_003751091.1 | 2 - 3 | 1897 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
