Assay Details
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078622.2 | NP_511177.2 | 7 - 8 | 810 | 65 | |
XM_006248444.2 | XP_006248506.1 | 7 - 8 | 963 | 65 | ||
XM_008768694.1 | XP_008766916.1 | 7 - 8 | 1161 | 65 | ||
GenBank mRNA | BC085713.1 | - | 7 - 8 | 808 | 7127059 | 65 |
L13245.1 | - | 6 - 7 | 707 | 65 | ||
M36071.1 | - | 7 - 8 | 807 | 65 | ||
U03490.1 | - | 7 - 8 | 772 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
