Assay Details
Gene Symbol: Gsk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032080.1 | NP_114469.1 | 9 - 10 | 1238 | 97 | |
XM_006248373.2 | XP_006248435.1 | 10 - 11 | 1983 | 97 | ||
XM_006248374.2 | XP_006248436.1 | 10 - 11 | 1983 | 97 | ||
XM_006248375.2 | XP_006248437.1 | 10 - 11 | 1983 | 97 | ||
XM_008768745.1 | XP_008766967.1 | 10 - 11 | 1983 | 97 | ||
GenBank mRNA | FQ231148.1 | - | 4 - 5 | 506 | 97 | |
FQ233111.1 | - | 4 - 5 | 506 | 97 | ||
X53428.1 | - | 9 - 10 | 1213 | 97 | ||
X73653.1 | - | 9 - 10 | 1238 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
re-entry into mitotic cell cycle
epithelial to mesenchymal transition
positive regulation of cell-matrix adhesion
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
protein export from nucleus
ER overload response
establishment or maintenance of cell polarity
axonogenesis
myoblast fusion
circadian rhythm
organ morphogenesis
response to lithium ion
negative regulation of cardiac muscle hypertrophy
positive regulation of peptidyl-threonine phosphorylation
regulation of neuron projection development
negative regulation of neuron projection development
negative regulation of neuron maturation
myotube differentiation
Wnt signaling pathway
phosphorylation
cell migration
peptidyl-serine phosphorylation
hippocampus development
establishment of cell polarity
negative regulation of protein complex assembly
positive regulation of protein complex assembly
negative regulation of protein binding
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of microtubule-based process
positive regulation of peptidyl-serine phosphorylation
protein localization to microtubule
intracellular signal transduction
cellular response to interleukin-3
response to drug
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAP kinase activity
hypermethylation of CpG island
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
fat cell differentiation
positive regulation of axon extension
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of protein export from nucleus
regulation of neuronal synaptic plasticity
negative regulation of dendrite morphogenesis
negative regulation of NFAT protein import into nucleus
canonical Wnt signaling pathway
superior temporal gyrus development
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
positive regulation of stem cell differentiation
RNA polymerase II transcription factor binding
p53 binding
protein kinase activity
protein serine/threonine kinase activity
integrin binding
protein binding
ATP binding
beta-catenin binding
transcription factor binding
kinase activity
protein kinase binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
ionotropic glutamate receptor binding
tau protein binding
tau-protein kinase activity
NF-kappaB binding
re-entry into mitotic cell cycle
epithelial to mesenchymal transition
positive regulation of cell-matrix adhesion
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
protein export from nucleus
ER overload response
establishment or maintenance of cell polarity
axonogenesis
myoblast fusion
circadian rhythm
organ morphogenesis
response to lithium ion
negative regulation of cardiac muscle hypertrophy
positive regulation of peptidyl-threonine phosphorylation
regulation of neuron projection development
negative regulation of neuron projection development
negative regulation of neuron maturation
myotube differentiation
Wnt signaling pathway
phosphorylation
cell migration
peptidyl-serine phosphorylation
hippocampus development
establishment of cell polarity
negative regulation of protein complex assembly
positive regulation of protein complex assembly
negative regulation of protein binding
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of microtubule-based process
positive regulation of peptidyl-serine phosphorylation
protein localization to microtubule
intracellular signal transduction
cellular response to interleukin-3
response to drug
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAP kinase activity
hypermethylation of CpG island
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
fat cell differentiation
positive regulation of axon extension
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of protein export from nucleus
regulation of neuronal synaptic plasticity
negative regulation of dendrite morphogenesis
negative regulation of NFAT protein import into nucleus
canonical Wnt signaling pathway
superior temporal gyrus development
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
positive regulation of stem cell differentiation
RNA polymerase II transcription factor binding
p53 binding
protein kinase activity
protein serine/threonine kinase activity
integrin binding
protein binding
ATP binding
beta-catenin binding
transcription factor binding
kinase activity
protein kinase binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
ionotropic glutamate receptor binding
tau protein binding
tau-protein kinase activity
NF-kappaB binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P620042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175040 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P620042 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P620042 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P620042 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P620042 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2582 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34106 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73503 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18043 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40756 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92447 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19341 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28773 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10159 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56665 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1688 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19146 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6623 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38124 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65950 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98538 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10754190 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754197 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754191 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754198 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754189 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754199 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754200 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10754188 | - | - |
Rat Genome 230 2.0 Array | 1370267_at | - | - |
Rat Genome 230 2.0 Array | 1370267_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370267_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370267_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100010032.rn.1 | TC1100001658.rn.1 | RTA TC |
Rat Expression Set 230 | 1370267_at | - | - |
Rat Expression Set 230 | 1370267_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754188 | - | - |
Rat Genome U34 Set | X73653_at | - | - |
Rat Genome U34 Set | X53428cds_s_at | - | - |
Rat Genome U34 Set | X73653_at | - | - |
Rat Genome U34 Set | X53428cds_s_at | - | - |
Rat Genome U34 Set | X53428cds_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349648 | - | - |
RatRef-12 Expression BeadChip | ILMN_49666 | - | - |
