Assay Details
Gene Symbol: Popdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199113.1 | NP_954544.1 | 3 - 4 | 1571 | 99 | |
GenBank mRNA | AY490241.1 | - | 3 - 4 | 1534 | 99 | |
BC061876.1 | - | 3 - 4 | 1571 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
