Assay Details
Gene Symbol: Nr1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052980.2 | NP_443212.1 | 2 - 3 | 437 | 152 | |
GenBank mRNA | AF151377.1 | - | 2 - 3 | 410 | 152 | |
BC091138.1 | - | 2 - 3 | 438 | 152 | ||
BC107449.1 | - | 1 - 2 | 199 | 152 | ||
CO575646.1 | - | 2 - 3 | 437 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
