Assay Details
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105879.1 | NP_001099349.1 | 11 - 12 | 2116 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P102280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98625 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102280 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102280 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P102280 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P302697 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83765 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25158 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23298 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3923 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10751256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751265 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751263 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751266 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751262 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10751255 | - | - |
Rat Genome 230 2.0 Array | 1395715_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395715_at | - | - |
Affymetrix HT Rat Focus Array | 1395715_at | - | - |
Affymetrix HT Rat Focus Array | 1395715_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100003373.rn.1 | TC1100000630.rn.1 | RTA TC |
Rat Expression Set 230 | 1395715_PM_at | - | - |
Rat Expression Set 230 | 1395715_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10751255 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21408968 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373980 | - | - |
