Assay Details
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134350.2 | NP_599177.2 | 9 - 10 | 1248 | 87 | |
XM_006248150.2 | XP_006248212.1 | 11 - 12 | 1392 | 87 | ||
XM_008768568.1 | XP_008766790.1 | 11 - 12 | 1326 | 87 | ||
GenBank mRNA | BC099808.1 | - | 9 - 10 | 1248 | 7938638 | 87 |
X52712.1 | - | 9 - 10 | 1323 | 87 | ||
X52713.1 | - | 8 - 9 | 1128 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
