Assay Details
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248353.2 | XP_006248415.1 | 15 - 16 | 2968 | 73 | |
XM_006248355.2 | XP_006248417.1 | 15 - 16 | 2965 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
