Assay Details
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751052.3 | XP_003751100.1 | 17 - 18 | 3241 | 101 | |
XM_006248353.2 | XP_006248415.1 | 17 - 18 | 3247 | 101 | ||
XM_006248354.2 | XP_006248416.1 | 17 - 18 | 3244 | 101 | ||
XM_006248355.2 | XP_006248417.1 | 17 - 18 | 3244 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
