Assay Details
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751052.3 | XP_003751100.1 | 7 - 8 | 1613 | 90 | |
XM_006248355.2 | XP_006248417.1 | 7 - 8 | 1613 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
