Assay Details
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751052.3 | XP_003751100.1 | 8 - 9 | 1779 | 56 | |
XM_006248353.2 | XP_006248415.1 | 8 - 9 | 1782 | 56 | ||
XM_006248354.2 | XP_006248416.1 | 8 - 9 | 1782 | 56 | ||
XM_006248355.2 | XP_006248417.1 | 8 - 9 | 1779 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
