Assay Details
Gene Symbol: Cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108525.1 | NP_001101995.1 | 6 - 7 | 702 | 72 | |
XM_006231526.1 | XP_006231588.1 | 6 - 7 | 691 | 72 | ||
XM_006231527.1 | XP_006231589.1 | 5 - 6 | 655 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
