Assay Details
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004261.1 | NP_001004261.1 | 3 - 4 | 247 | 103 | |
XM_006231394.2 | XP_006231456.1 | 3 - 4 | 518 | 103 | ||
XM_006231396.2 | XP_006231458.1 | 3 - 4 | 519 | 103 | ||
XM_008760424.1 | XP_008758646.1 | 3 - 4 | 521 | 103 | ||
XM_008760425.1 | XP_008758647.1 | 3 - 4 | 284 | 103 | ||
GenBank mRNA | BC079368.1 | - | 3 - 4 | 247 | 7109628 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
