Assay Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107448.2 | NP_001100918.1 | 4 - 5 | 522 | 131 | |
XM_006231673.2 | XP_006231735.1 | 4 - 5 | 1338 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
