Assay Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107448.2 | NP_001100918.1 | 7 - 8 | 813 | 65 | |
XM_006231673.2 | XP_006231735.1 | 7 - 8 | 1629 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P316415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144245 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P316415 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P316415 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P316415 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P316415 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95101 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10731150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10731152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10731148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10731151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10731149 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10731147 | - | - |
Rat Genome 230 2.0 Array | 1394566_at | - | - |
Rat Genome 230 2.0 Array | 1394566_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392663_at | - | - |
Rat Genome 230 2.0 Array | 1392663_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394566_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392663_at | - | - |
Affymetrix HT Rat Focus Array | 1394566_at | - | - |
Affymetrix HT Rat Focus Array | 1392663_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100064282.rn.1 | TC0100007984.rn.1 | RTA TC |
Rat Expression Set 230 | 1394566_PM_at | - | - |
Rat Expression Set 230 | 1394566_at | - | - |
Rat Expression Set 230 | 1392663_PM_at | - | - |
Rat Expression Set 230 | 1392663_at | - | - |
Rat Genome U34 Set | rc_AI179925_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20903883 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357698 | - | - |
