Assay Details
Gene Symbol: Prdx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060567.1 | - | 7 - 8 | 719 | 6922338 | 119 |
FQ212826.1 | - | 6 - 7 | 728 | 119 | ||
FQ219655.1 | - | 6 - 7 | 745 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
